N-[2-(Isopentyloxy)benzyl]-4-methylaniline

Supplier: Matrix Scientific

031572-500MG
101831-970EA 306.15 USD
101831-970
N-[2-(Isopentyloxy)benzyl]-4-methylaniline
N-[2-(Isopentyloxy)benzyl]-4-methylaniline

Hazard: Irritant

Formula: C₁₉H₂₅NO
MDL Number: MFCD10687642
CAS Number: 1040688-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR