N-(4-Methylphenyl)-N-[2-(1-naphthyl)ethyl]amine

Supplier: Matrix Scientific

031570-500MG
101831-966EA 306.15 USD
101831-966
N-(4-Methylphenyl)-N-[2-(1-naphthyl)ethyl]amine
N-(4-Methylphenyl)-N-[2-(1-naphthyl)ethyl]amine

Hazard: Irritant

Formula: C₁₉H₁₉N
MDL Number: MFCD10687640
CAS Number: 1040688-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR