N-(Cyclopropylmethyl)-4-methylaniline

Supplier: Matrix Scientific

031569-500MG
101831-964EA 306.15 USD
101831-964
N-(Cyclopropylmethyl)-4-methylaniline
N-(Cyclopropylmethyl)-4-methylaniline

Hazard: Irritant

Formula: C₁₁H₁₅N
MDL Number: MFCD03211292
CAS Number: 356539-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR