4-Methyl-N-[2-(4-methylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031568-500MG
101831-962EA 306.15 USD
101831-962
4-Methyl-N-[2-(4-methylphenoxy)ethyl]aniline
4-Methyl-N-[2-(4-methylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₆H₁₉NO
MDL Number: MFCD00086444
CAS Number: 200806-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR