4-Methyl-N-(3-methylbenzyl)aniline

Supplier: Matrix Scientific

031567-500MG
101831-960EA 306.15 USD
101831-960
4-Methyl-N-(3-methylbenzyl)aniline
4-Methyl-N-(3-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₅H₁₇N
MDL Number: MFCD03210609
CAS Number: 33863-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR