4-Methyl-N-(3-phenylpropyl)aniline

Supplier: Matrix Scientific

031566-500MG
101831-958EA 306.15 USD
101831-958
4-Methyl-N-(3-phenylpropyl)aniline
4-Methyl-N-(3-phenylpropyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₉N
MDL Number: MFCD01679498
CAS Number: 63980-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR