2,5-Dimethoxy-N-[3-(phenethyloxy)benzyl]aniline

Supplier: Matrix Scientific

031563-500MG
101831-952EA 306.15 USD
101831-952
2,5-Dimethoxy-N-[3-(phenethyloxy)benzyl]aniline
2,5-Dimethoxy-N-[3-(phenethyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₃
MDL Number: MFCD10687637
CAS Number: 1040688-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR