N-[2-(Isopentyloxy)benzyl]-2,5-dimethoxyaniline

Supplier: Matrix Scientific

031561-500MG
101831-948EA 306.15 USD
101831-948
N-[2-(Isopentyloxy)benzyl]-2,5-dimethoxyaniline
N-[2-(Isopentyloxy)benzyl]-2,5-dimethoxyaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₃
MDL Number: MFCD10687635
CAS Number: 1040688-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR