2,5-Dimethoxy-N-[2-(4-methylphenoxy)butyl]aniline

Supplier: Matrix Scientific

031559-500MG
101831-944EA 306.15 USD
101831-944
2,5-Dimethoxy-N-[2-(4-methylphenoxy)butyl]aniline
2,5-Dimethoxy-N-[2-(4-methylphenoxy)butyl]aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₃
MDL Number: MFCD10687633
CAS Number: 1040688-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR