2,5-Dimethoxy-N-(4-methoxyphenethyl)aniline

Supplier: Matrix Scientific

031557-500MG
101831-940EA 306.15 USD
101831-940
2,5-Dimethoxy-N-(4-methoxyphenethyl)aniline
2,5-Dimethoxy-N-(4-methoxyphenethyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₁NO₃
MDL Number: MFCD03211149
CAS Number: 356537-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR