2,5-Dimethoxy-N-(2-thienylmethyl)aniline

Supplier: Matrix Scientific

031556-500MG
101831-938EA 306.15 USD
101831-938
2,5-Dimethoxy-N-(2-thienylmethyl)aniline
2,5-Dimethoxy-N-(2-thienylmethyl)aniline

Hazard: Irritant

Formula: C₁₃H₁₅NO₂S
MDL Number: MFCD10687632
CAS Number: 1021029-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR