3-Chloro-2-methyl-N-(2-methylphenethyl)aniline

Supplier: Matrix Scientific

031554-500MG
101831-934EA 306.15 USD
101831-934
3-Chloro-2-methyl-N-(2-methylphenethyl)aniline
3-Chloro-2-methyl-N-(2-methylphenethyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₈ClN
MDL Number: MFCD10687630
CAS Number: 1040688-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR