3-Chloro-N-[4-(isopentyloxy)benzyl]-2-methylaniline

Supplier: Matrix Scientific

031553-500MG
101831-932EA 306.15 USD
101831-932
3-Chloro-N-[4-(isopentyloxy)benzyl]-2-methylaniline
3-Chloro-N-[4-(isopentyloxy)benzyl]-2-methylaniline

Hazard: Irritant

Formula: C₁₉H₂₄ClNO
MDL Number: MFCD10687629
CAS Number: 1040688-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR