3-Chloro-2-methyl-N-[4-(pentyloxy)benzyl]aniline

Supplier: Matrix Scientific

031551-500MG
101831-928EA 306.15 USD
101831-928
3-Chloro-2-methyl-N-[4-(pentyloxy)benzyl]aniline
3-Chloro-2-methyl-N-[4-(pentyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₁₉H₂₄ClNO
MDL Number: MFCD10687627
CAS Number: 1040688-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR