3-Chloro-2-methyl-N-(4-methylbenzyl)aniline

Supplier: Matrix Scientific

031547-500MG
101831-920EA 306.15 USD
101831-920
3-Chloro-2-methyl-N-(4-methylbenzyl)aniline
3-Chloro-2-methyl-N-(4-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₅H₁₆ClN
MDL Number: MFCD03210667
CAS Number: 356530-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR