3-Chloro-N-(4-methoxybenzyl)-2-methylaniline

Supplier: Matrix Scientific

031545-500MG
101831-916EA 306.15 USD
101831-916
3-Chloro-N-(4-methoxybenzyl)-2-methylaniline
3-Chloro-N-(4-methoxybenzyl)-2-methylaniline

Hazard: Irritant

Formula: C₁₅H₁₆ClNO
MDL Number: MFCD01826289
CAS Number: 356537-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR