3-Chloro-N-(4-fluorobenzyl)-2-methylaniline

Supplier: Matrix Scientific

031544-500MG
101831-914EA 306.15 USD
101831-914
3-Chloro-N-(4-fluorobenzyl)-2-methylaniline
3-Chloro-N-(4-fluorobenzyl)-2-methylaniline

Hazard: Irritant

Formula: C₁₄H₁₃ClFN
MDL Number: MFCD03210775
CAS Number: 356531-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR