3-(2-Oxo-2H-pyridin-1-yl)propionic acid hydrobromide

Supplier: Matrix Scientific

034665-500MG
101838-106EA 307.69 USD
101838-106
3-(2-Oxo-2H-pyridin-1-yl)propionic acid hydrobromide
3-(2-Oxo-2H-pyridin-1-yl)propionic acid hydrobromide

Hazard: Irritant

Formula: C₈H₁₀BrNO₃
MDL Number: MFCD11506643
CAS Number: 878775-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR