1-(1,1-Dioxo-tetrahydro-1λ*6*-thiophen-3-yl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

034658-500MG
101838-092EA 307.69 USD
101838-092
1-(1,1-Dioxo-tetrahydro-1λ*6*-thiophen-3-yl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
1-(1,1-Dioxo-tetrahydro-1lambda*6*-thiophen-3-yl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₄S
MW: 258.3 g/mol
MDL Number: MFCD09863869
CAS Number: 1134555-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR