2-(2,4-Difluorophenyl)-2-morpholin-4-ylethylamine hydrochloride

Supplier: Matrix Scientific

034652-500MG
101838-080EA 183.08 USD
101838-080
2-(2,4-Difluorophenyl)-2-morpholin-4-ylethylamine hydrochloride
2-(2,4-Difluorophenyl)-2-morpholin-4-ylethylamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClF₂N₂O
MDL Number: MFCD11506641
CAS Number: 1185293-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR