1-(5-Amino-2-methoxyphenyl)pyrrolidin-2-one

Supplier: Matrix Scientific

034647-500MG
101838-070EA 204.62 USD
101838-070
1-(5-Amino-2-methoxyphenyl)pyrrolidin-2-one
1-(5-Amino-2-methoxyphenyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD09732748
CAS Number: 69131-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR