2-Isopropyl-4-methylpyrimidine-5-carboxylic acid

Supplier: Matrix Scientific

034643-500MG
101838-062EA 252.31 USD
101838-062
2-Isopropyl-4-methylpyrimidine-5-carboxylic acid
2-Isopropyl-4-methylpyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD09750210
CAS Number: 954233-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR