2-(3-Methoxyphenyl)-2-methylpropylamine

Supplier: Matrix Scientific

034642-500MG
101838-060EA 384.62 USD
101838-060
2-(3-Methoxyphenyl)-2-methylpropylamine
2-(3-Methoxyphenyl)-2-methylpropylamine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD09414706
CAS Number: 60812-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR