2-Methyl-6-(trifluoromethyl)nicotinic acid

Supplier: Matrix Scientific

034623-500MG
101838-022EA 40 USD
101838-022
2-Methyl-6-(trifluoromethyl)nicotinic acid
2-Methyl-6-(trifluoromethyl)nicotinic acid

Hazard: Irritant

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
MDL Number: MFCD01311987
CAS Number: 261635-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR