1-Cyclohexylmethyl-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

034602-500MG
101837-980EA 384.62 USD
101837-980
1-Cyclohexylmethyl-5-oxopyrrolidine-3-carboxylic acid
1-Cyclohexylmethyl-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD09055063
CAS Number: 304859-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR