2-Isopropylpyrimidine-5-carboxylic acid

Supplier: Matrix Scientific

034589-500MG
101837-954EA 384.62 USD
101837-954
2-Isopropylpyrimidine-5-carboxylic acid
2-Isopropylpyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD09054857
CAS Number: 927803-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR