3-[(4-Fluorophenyl)methylamino]propionic acid

Supplier: Matrix Scientific

034576-500MG
101837-928EA 307.69 USD
101837-928
3-[(4-Fluorophenyl)methylamino]propionic acid
3-[(4-Fluorophenyl)methylamino]propionic acid

Hazard: Irritant

Formula: C₁₀H₁₂FNO₂
MDL Number: MFCD09755076
CAS Number: 944886-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR