3-(Methyl-m-tolylamino)propionic acid

Supplier: Matrix Scientific

034571-500MG
101837-918EA 384.62 USD
101837-918
3-(Methyl-m-tolylamino)propionic acid
3-(Methyl-m-tolylamino)propionic acid

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD09755083
CAS Number: 944887-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR