3-(5-Ethyl[1,3,4]oxadiazol-2-yl)phenylamine

Supplier: Matrix Scientific

034568-500MG
101837-912EA 461.54 USD
101837-912
3-(5-Ethyl[1,3,4]oxadiazol-2-yl)phenylamine
3-(5-Ethyl[1,3,4]oxadiazol-2-yl)phenylamine

Hazard: Irritant

Formula: C₁₀H₁₁N₃O
MDL Number: MFCD11052290
CAS Number: 954329-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR