3-Morpholin-4-yl-3-phenylpropionic acid

Supplier: Matrix Scientific

034559-500MG
101837-894EA 484.62 USD
101837-894
3-Morpholin-4-yl-3-phenylpropionic acid
3-Morpholin-4-yl-3-phenylpropionic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MDL Number: MFCD09035612
CAS Number: 937652-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR