MATR034557

034557-500MG
101837-890EA 326.92 USD
101837-890
MATR034557
CAS: 85301-38-8
MDL Number: MFCD08753718
Molecular Formula: C6H7N3O3
Molecular Weight: 169,14
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR