(4,5,6-Trimethyl-2-oxo-2H-pyrimidin-1-yl)acetic acid hydrochloride

Supplier: Matrix Scientific

034556-500MG
101837-888EA 183.08 USD
101837-888
(4,5,6-Trimethyl-2-oxo-2H-pyrimidin-1-yl)acetic acid hydrochloride
(4,5,6-Trimethyl-2-oxo-2H-pyrimidin-1-yl)acetic acid hydrochloride

Hazard: Irritant

Formula: C₉H₁₃ClN₂O₃
MDL Number: MFCD11506627
CAS Number: 1185294-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR