1-Ethyl-1H-benzoimidazole-5-carboxylic acid

Supplier: Matrix Scientific

034549-500MG
101837-874EA 384.62 USD
101837-874
1-Ethyl-1H-benzoimidazole-5-carboxylic acid
1-Ethyl-1H-benzoimidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD11052244
CAS Number: 943110-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR