3-(3-Pyrrolidin-1-ylpropoxy)benzoic acid hydrochloride

Supplier: Matrix Scientific

034544-500MG
101837-864EA 384.62 USD
101837-864
3-(3-Pyrrolidin-1-ylpropoxy)benzoic acid hydrochloride
3-(3-Pyrrolidin-1-ylpropoxy)benzoic acid hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD11506625
CAS Number: 1185303-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR