4-Methyl-3-pyrrolidin-1-ylbenzoic acid

Supplier: Matrix Scientific

034543-500MG
101837-862EA 384.62 USD
101837-862
4-Methyl-3-pyrrolidin-1-ylbenzoic acid
4-Methyl-3-pyrrolidin-1-ylbenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₂
MW: 205.26 g/mol
Melting Pt: 163…165 °C
MDL Number: MFCD06740067
CAS Number: 107946-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR