6-Oxo-1,6-dihydropyrimidine-5-carboxylic acid

Supplier: Matrix Scientific

034532-500MG
101837-840EA 75.38 USD
101837-840
6-Oxo-1,6-dihydropyrimidine-5-carboxylic acid
6-Oxo-1,6-dihydropyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₅H₄N₂O₃
MW: 140.1000 g/mol
MDL Number: MFCD08692965
CAS Number: 65754-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR