4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid

Supplier: Matrix Scientific

Synonyms: 4-Methyl-2-(pyridin-4-yl)thiazole-5-carboxylic acid

034515-500MG
101837-806EA 307.69 USD
101837-806
4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid
4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂S
MW: 220.2477 g/mol
MDL Number: MFCD00171757
CAS Number: 144060-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR