2-(3-Methoxyphenoxy)-2-methylpropionic acid sodium salt

Supplier: Matrix Scientific

034513-500MG
101837-802EA 307.69 USD
101837-802
2-(3-Methoxyphenoxy)-2-methylpropionic acid sodium salt
2-(3-Methoxyphenoxy)-2-methylpropionic acid sodium salt

Hazard: Irritant

Formula: C₁₁H₁₃NaO₄
MDL Number: MFCD11506616
CAS Number: 1185090-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR