3-(2-Oxo-2-pyrrolidin-1-ylethoxy)benzoic acid

Supplier: Matrix Scientific

034474-500MG
101837-726EA 307.69 USD
101837-726
3-(2-Oxo-2-pyrrolidin-1-ylethoxy)benzoic acid
3-(2-Oxo-2-pyrrolidin-1-ylethoxy)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD09221904
CAS Number: 923173-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR