2-(5-Fluoro-1-methyl-1H-indol-3-yl)-1-methylethylamine hydrochloride

Supplier: Matrix Scientific

034467-500MG
101837-712EA 538.46 USD
101837-712
2-(5-Fluoro-1-methyl-1H-indol-3-yl)-1-methylethylamine hydrochloride
2-(5-Fluoro-1-methyl-1H-indol-3-yl)-1-methylethylamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClFN₂
MDL Number: MFCD08456048
CAS Number: 1185301-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR