Diethylsulfamoyl chloride

Supplier: Matrix Scientific

034446-500MG
101837-670EA 307.69 USD
101837-670
Diethylsulfamoyl chloride
Diethylsulfamoyl chloride

Hazard: Irritant

Formula: C₄H₁₀ClNO₂S
MDL Number: MFCD03249508
CAS Number: 20588-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR