[1-(4-Fluorophenyl)propyl]methylamine

Supplier: Matrix Scientific

034441-500MG
101837-660EA 307.69 USD
101837-660
[1-(4-Fluorophenyl)propyl]methylamine
[1-(4-Fluorophenyl)propyl]methylamine

Hazard: Irritant

Formula: C₁₀H₁₄FN
MDL Number: MFCD09043960
CAS Number: 926262-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR