3-(5-Chloro-1-methyl-1H-benzoimidazol-2-yl)propionic acid hydrochloride

Supplier: Matrix Scientific

034433-500MG
101837-644EA 307.69 USD
101837-644
3-(5-Chloro-1-methyl-1H-benzoimidazol-2-yl)propionic acid hydrochloride
3-(5-Chloro-1-methyl-1H-benzoimidazol-2-yl)propionic acid hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₂Cl₂N₂O₂
MDL Number: MFCD18385064
CAS Number: 1354543-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR