3-Ethyl-1,5-dimethyl-3,7-diazabicyclo[3.3.1]nonane

Supplier: Matrix Scientific

034410-500MG
101837-598EA 484.62 USD
101837-598
3-Ethyl-1,5-dimethyl-3,7-diazabicyclo[3.3.1]nonane
3-Ethyl-1,5-dimethyl-3,7-diazabicyclo[3.3.1]nonane

Hazard: Irritant

Formula: C₁₁H₂₂N₂
MW: 182.31 g/mol
MDL Number: MFCD11052215
CAS Number: 917202-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR