1-(4-Aminophenyl)-2-(4-methylpiperazin-1-yl)ethanol

Supplier: Matrix Scientific

034378-500MG
101837-534EA 384.62 USD
101837-534
1-(4-Aminophenyl)-2-(4-methylpiperazin-1-yl)ethanol
1-(4-Aminophenyl)-2-(4-methylpiperazin-1-yl)ethanol

Hazard: Irritant

Formula: C₁₃H₂₁N₃O
MDL Number: MFCD08690214
CAS Number: 919021-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR