2-(2-Ethylimidazol-1-yl)propionic acid hydrochloride

Supplier: Matrix Scientific

034365-500MG
101837-510EA 227.69 USD
101837-510
2-(2-Ethylimidazol-1-yl)propionic acid hydrochloride
2-(2-Ethylimidazol-1-yl)propionic acid hydrochloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂O₂
MDL Number: MFCD11506601
CAS Number: 1219346-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR