5-Morpholin-4-yl-2-propoxy-benzoic acid hydrochloride

Supplier: Matrix Scientific

034364-500MG
101837-508EA 307.69 USD
101837-508
5-Morpholin-4-yl-2-propoxy-benzoic acid hydrochloride
5-Morpholin-4-yl-2-propoxy-benzoic acid hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₄
MDL Number: MFCD11506600
CAS Number: 1011366-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR