3-Cyclobutylpropionic acid

Supplier: Matrix Scientific

034357-500MG
101837-494EA 453.85 USD
101837-494
3-Cyclobutylpropionic acid
3-Cyclobutylpropionic acid

Hazard: Irritant

Formula: C₇H₁₂O₂
MDL Number: MFCD08061939
CAS Number: 4415-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR