2-(4-Fluorophenyl)-2-methylpropylamine

Supplier: Matrix Scientific

034344-500MG
101837-468EA 307.69 USD
101837-468
2-(4-Fluorophenyl)-2-methylpropylamine
2-(4-Fluorophenyl)-2-methylpropylamine

Hazard: Irritant

Formula: C₁₀H₁₄FN
MDL Number: MFCD09904312
CAS Number: 40377-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR