(2-Phenyloxazol-4-yl)acetic acid

Supplier: Matrix Scientific

034328-500MG
101837-436EA 576.92 USD
101837-436
(2-Phenyloxazol-4-yl)acetic acid
(2-Phenyloxazol-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₉NO₃
MDL Number: MFCD07838437
CAS Number: 22086-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR